Mettler Toledo Expert Pro pH Electrode; BNC with 3.9 ft Cable

Representative image only.

Mettler Toledo Expert Pro pH Electrode; BNC with 3.9 ft Cable

Mettler Toledo | Mfr: 51343101| Item: 3590006

Expertly-engineered probes to cover a wide range of applications

$766.16CAD / Each

$766.16CAD / Each

Buy It Together

Current Item

Mettler Toledo Expert Pro pH Electrode; BNC with 3.9 ft Cable
Mettler Toledo Expert Pro pH Electrode; BNC with 3.9 ft Cable
3590006

$766.16CAD / Each
+ + + + + =
Total Price 3961.93 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ARGENTHAL™ reference system⌡Open junction works well for dirty and complex samples

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

Open junction provides easy cleaning and clog-free measurements. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

Specifications
  • ApplicationsGeneral-use
  • Electrode Body MaterialPEEK
  • Reference JunctionOpen
  • Reference ElectrolyteNon-refillable Xerolyt® polymer
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • GuardYes
  • ATC ConnectorRCA (cinch)
  • ATC ElementNTC 30 kΩ
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1.2
  • DescriptionpH Electrode; BNC with 3.9 ft Cable
Reviews
Q&A