Mettler Toledo Expert pH Electrode; S7
Representative image only.
Mettler Toledo Expert pH Electrode; S7
Mettler Toledo | Mfr: 51343100| Item: 3590005
Expertly-engineered probes to cover a wide range of applications
$526.00CAD / Each
$526.00CAD / Each
Buy It Together
Current Item
Mettler Toledo Expert pH Electrode; S7
Mettler Toledo Expert pH Electrode; S7
3590005
$526.00CAD / Each
+ + + + + =
Total Price 3721.77 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Cable not included⌡ARGENTHAL™ reference system⌡Open junction works well for dirty and complex samples
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available.⌡Open junction provides easy cleaning and clog-free measurements. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.
Specifications
  • ApplicationsGeneral-use
  • Electrode Body MaterialPEEK
  • Reference JunctionOpen
  • Reference ElectrolyteNon-refillable Xerolyt® polymer
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • GuardYes
  • ATC Element
  • ConnectorS7
  • Diameter (mm)12
  • Length (mm)120
  • DescriptionpH Electrode; S7