Mettler Toledo Expert Go-ISM pH Electrode; BNC with 32.8 ft Cable

Representative image only.

Mettler Toledo Expert Go-ISM pH Electrode; BNC with 32.8 ft Cable

Mettler Toledo | Mfr: 51344104| Item: 3590011

Expertly-engineered probes to cover a wide range of applications

$1,156.61CAD / Each

$1,156.61CAD / Each

Buy It Together

Current Item

Mettler Toledo Expert Go-ISM pH Electrode; BNC with 32.8 ft Cable
Mettler Toledo Expert Go-ISM pH Electrode; BNC with 32.8 ft Cable
3590011

$1,156.61CAD / Each
+ + + + + =
Total Price 4352.38 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ISM® - Intelligent Sensor Management⌡ARGENTHAL™ reference system⌡Open junction works well for dirty and complex samples

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ISM technology stores calibration data in the sensor and automatically transfer it to the meter. Open junction provides easy cleaning and clog-free measurements. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

Specifications
  • ApplicationsGeneral-use
  • Electrode Body MaterialPEEK
  • Reference JunctionTwo open junctions
  • Reference ElectrolyteNon-refillable Xerolyt® polymer
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • GuardYes
  • ATC ConnectorRCA (cinch)
  • ATC ElementNTC 30 kΩ
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)10
  • DescriptionpH Electrode; BNC with 32.8 ft Cable
Reviews
Q&A