Mettler Toledo Easy BNC pH Electrode; BNC with 3.9 ft Cable

Representative image only.

Mettler Toledo Easy BNC pH Electrode; BNC with 3.9 ft Cable

Mettler Toledo | Mfr: 51343011| Item: 3590041

Expertly-engineered probes to cover a wide range of applications

Call For Price

Call For Price

Buy It Together

Current Item

Mettler Toledo Easy BNC pH Electrode; BNC with 3.9 ft Cable
Mettler Toledo Easy BNC pH Electrode; BNC with 3.9 ft Cable
3590041

Call For Price
+ + + + + =
Total Price 2834.50 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

Specifications
  • ApplicationsGeneral-use
  • Electrode Body MaterialPolysulfone (PS)
  • Reference JunctionCeramic
  • Reference ElectrolyteNon-refillable gel
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • GuardYes
  • ATC Element
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1.2
  • DescriptionpH Electrode; BNC with 3.9 ft Cable
Reviews
Q&A