Mettler Toledo cSmartCal24 Moisture Analyzer Reference Substance with Certificate; 24 Tests
Representative image only.
Mettler Toledo cSmartCal24 Moisture Analyzer Reference Substance with Certificate; 24 Tests
Mettler Toledo | Mfr: 30005791| Item: 1133629
Quick and Easy Performance Test for Moisture Analyzers
Call For Price
Call For Price
Product Description
Includes
24 test sachets and certificate of analysis.
Key Features
  • Indicates incorrect performance immediately
  • Test temperatures include 70, 100, 130, 160°C (158, 212, 266, 320°F)
  • Certified by an independent institute and includes a certificate of analysis
  • Three month warranty
More About this Item
SmartCal verifies the performance of your moisture analyzer in a 10-minute test, which is carried out in the same easy way as a regular measurement. Regular testing with SmartCal lets you provides documented evidence of the unit's performance over time. Incorrect performance is immediately indicated. The test instantly indicates adverse influences from the heating or weighing units. If the test is passed, you can trust your results.
3 Month Warranty
Specifications
  • Accessory TypeFor use with Mettler Toledo Halogen Moisture Analyzer
  • DescriptionSmartCal Moisture Analyzer Reference Substance with Certificate; 24 Tests