Mettler Toledo 30988512 EVA C3 Coulometric Karl Fischer Titrator, Generator Electrode without Diaphragm

Representative image only.

Mettler Toledo 30988512 EVA C3 Coulometric Karl Fischer Titrator, Generator Electrode without Diaphragm

Mettler Toledo | Mfr: 30988512| Item: 3333224

Accurately and efficiently determine the water content for ultra-low (1 ppm to 5%) samples

$23,338.50CAD / Each

$23,338.50CAD / Each

Buy It Together

Current Item

Mettler Toledo 30988512 EVA C3 Coulometric Karl Fischer Titrator, Generator Electrode without Diaphragm
Mettler Toledo 30988512 EVA C3 Coulometric Karl Fischer Titrator, Generator Electrode without Diaphragm
3333224

$23,338.50CAD / Each
+ =
Total Price 24745.59 ADD 2 ITEMS TO CART
Product Description

Includes

Generator electrode without diaphragm, generator electrode cable, titration kit, solvent pump dPump KF kit, dSens M143 sensor kit, titrator cover, touchscreen protective cover, terminal cable, dSens dVP4-T cable, external power supply, and power cable.

Key Features

  • Fast repeatable measurements without sacrificing accuracy
  • Electrochemically generated iodine helps measure extremely low water levels
  • Automatic solvent exchange of reduces contact with chemicals
  • Sealed, easy-to-clean hybrid cell streamlines cleaning, reducing the risk of contamination
  • Easy-to-understand One Click™ touchscreen user interface
  • Save shortcuts to minimize errors and save time
  • Flexible data export via USB ports or Ethernet
  • Use for determining water content of simple liquids, soluble solids, and insoluble samples
  • Comes with a generator electrode without diaphragm
  • Compatible with InMotion KF Six, KF Flex, or KF Pro autosampler
  • 1 year warranty

More About this Item

Ensure the quality of your product by accurately determining the water content in your sample. The EVA Karl Fischer titrators simplify sample handling and eliminate the risk of human error, delivering highly reproducible and accurate results within a matter of minutes. Coulometric titrators analyze liquids, soluble solids, and insoluble solids (EVA C3 only) with water content between 1 ppm to 5% with accuracy and efficiency.

A combination of the Fast Forecasting Amperometric (FFA™) control algorithm, a fast-response digital sensor, and state-of-the-art dispensing technology ensure fast and accurate results. Coulometric Karl Fischer titrators generate iodine electrochemically from iodide, allowing for precise quantification of low water levels. Use the large color touchscreen to operate the simple-to-use interface to customize shortcuts and simply daily tasks.

Ensure user safety with the solvent manager. It is fully controlled by the titrator, and offers filling, draining, and solvent exchange without additional user interaction. For secure titrator operation, designate user groups to allow full or limited access. Easily export data in CSV, PDF, or XML files using a memory stick, connect to a printer via the USB port, or network export via Ethernet.

This model includes a generator electrode without diaphragm. Electrode without diaphragm is easier to clean than an electrode with diaphragm. It performs automatic exchange of anolyte and has a slightly lower accuracy for samples with low amounts of water.

Conformite Europeenne1 Year Warranty
Specifications
  • Units%, mg, mg/g, mg/kg, mg/mL, ppm, g/100g, g/100mL, g/kg, g/mL, µg/L, µg/mL, µg
  • Display Type7" TFT IPS color display with capacitive multitouch screen
  • ApplicationsLiquids, soluble solids, and insoluble solids
  • Display Resolution1024 x 600 pixels
  • Min Temperature (° C)18
  • Max Temperature (DegreeC)28
  • OutputUSB (2) and Ethernet
  • Data LoggingYes
  • Special FeaturesGenerator electrode without diaphragm
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Length (in)7
  • Width (in)5 5/16
  • Height (in)7 1/4
  • Length (cm)17.7
  • Width (cm)13.5
  • Height (cm)18.5
  • DescriptionEVA C3 Coulometric Karl Fischer Titrator, Generator Electrode without Diaphragm
  • Warranty1 year
Reviews
Q&A