Meta Fluid-Resistant Antistatic Lab Coat, X-Large
Representative image only.
Meta Fluid-Resistant Antistatic Lab Coat, X-Large
Meta | Mfr: 11653-011-XL| Item: 8646843
Features carbon fiber for static dissipation
$147.34CAD / Each
BUY MORE & SAVE
  • 1-5$147.34 CAD / Each
  • 6+$132.61 CAD / Each
$147.34CAD / Each
BUY MORE & SAVE
  • 1-5$147.34 CAD / Each
  • 6+$132.61 CAD / Each
Product Description
More About this Item
Coats are made of 99% textured polyester for increased fluid resistance and 1% carbon fiber for static dissipation. The carbon fiber shows as pin-striping on the coat. Coats also feature five-button front, extra button closure at the neck for full coverage, three pockets, back pleats for extra movement, and knit cuffs for better wrist coverage.
Specifications
  • Length (in)40
  • Length (cm)101.6
  • MaterialCarbon Fiber;Polyester
  • SizeX-Large
  • ColorWhite
  • DescriptionFluid-Resistant Antistatic Lab Coat, X-Large