Meta Fluid-Resistant Antistatic Lab Coat, Small
Representative image only.
Meta Fluid-Resistant Antistatic Lab Coat, Small
Meta | Mfr: 11653-011-S| Item: 8646840
Features carbon fiber for static dissipation
$149.18CAD / Each
BUY MORE & SAVE
  • 1-5$149.18 CAD / Each
  • 6+$134.26 CAD / Each
$149.18CAD / Each
BUY MORE & SAVE
  • 1-5$149.18 CAD / Each
  • 6+$134.26 CAD / Each
Product Description
More About this Item
Coats are made of 99% textured polyester for increased fluid resistance and 1% carbon fiber for static dissipation. The carbon fiber shows as pin-striping on the coat. Coats also feature five-button front, extra button closure at the neck for full coverage, three pockets, back pleats for extra movement, and knit cuffs for better wrist coverage.
Specifications
  • Length (in)40
  • Length (cm)101.6
  • MaterialPolyester;Carbon Fiber
  • SizeSmall
  • ColorWhite
  • DescriptionFluid-Resistant Antistatic Lab Coat, Small