MediaKap ME2M-50B-03S Hollow Fiber Media Filters, 25-50 L, 1/4-3/8" ID, Bell; 3/Pk

Representative image only.

MediaKap ME2M-50B-03S Hollow Fiber Media Filters, 25-50 L, 1/4-3/8" ID, Bell; 3/Pk

MediaKap | Mfr: ME2M-50B-03S| Item: 2951008

Sterilize and clarify cell culture media and aqueous solutions quickly and efficiently

$732.94CAD / Box of 3

$732.94CAD / Box of 3
Product Description

Key Features

  • Filters may be used as pressure filters with peristaltic pumps, pressure vessels, or repeating syringes
  • PVC filling bell in models maintains sterility downstream

More About this Item

MediaKap® Hollow Fiber Media Filter provides biocompatibility, low extractables, and high flow rates for serum-free media, and aqueous solutions. Polypropylene autovent prevents air locking. Naturally hydrophilic, 0.2 µm microporous membrane.

Sterile
Specifications
  • Membrane MaterialMixed Cellulose Ester
  • Pore Size (micron)0.2
  • Housing MaterialPolycarbonate
  • SterileYes
  • Connections1/4" to 3/8" hose barb
  • Effective Filtration Area (cm²)440
  • Surface Area (m²)0.044
  • Typical Water Flow Rate2000 mL/min at 10 psi
  • Max Operating Temperature (° C)83
  • Max Operating Temperature (° F)182
  • Max Pressure (PSI)50
  • Max Pressure3.45
  • Min Throughput (Liters)25
  • Max Throughput (Liters)50
  • Min Throughput (mL)25000
  • Max Throughput (mL)50000
  • DescriptionHollow Fiber Media Filters, 25-50 L, 1/4-3/8" ID, Bell; 3/Pk
Reviews
Q&A