MediaKap ME2M-10B-12S Hollow Fiber Media Filters, 1000 mL/min Flow Rate

Representative image only.

MediaKap ME2M-10B-12S Hollow Fiber Media Filters, 1000 mL/min Flow Rate

MediaKap | Mfr: ME2M-10B-12S| Item: 2951014

Sterilize and clarify cell culture media and aqueous solutions quickly and efficiently

$1,471.25CAD / Box of 12

$1,471.25CAD / Box of 12
Product Description

Key Features

  • Filters may be used as pressure filters with peristaltic pumps, pressure vessels, or repeating syringes
  • PVC filling bell in models maintains sterility downstream
  • 1 year warranty

More About this Item

MediaKap® Hollow Fiber Media Filter provides biocompatibility, low extractables, and high flow rates for serum-free media, and aqueous solutions. Polypropylene autovent prevents air locking. Naturally hydrophilic, 0.2 µm microporous membrane.

Sterile1 Year Warranty
Specifications
  • Membrane MaterialMixed Cellulose Ester
  • Pore Size (micron)0.2
  • Housing MaterialPolycarbonate
  • SterileYes
  • Connections1/4" hose barb
  • Effective Filtration Area (cm²)100
  • Surface Area (m²)0.01
  • Typical Water Flow Rate1000 mL/min at 10 psi
  • Max Operating Temperature (° C)83
  • Max Operating Temperature (° F)182
  • Max Pressure (PSI)50
  • Max Pressure3.45
  • Min Throughput (Liters)5
  • Max Throughput (Liters)10
  • Min Throughput (mL)5000
  • Max Throughput (mL)10000
  • DescriptionHollow Fiber Media Filters, 1000 mL/min Flow Rate
  • Warranty1 year
Reviews
Q&A