MediaKap ME2M-050-18S Hollow Fiber Media Filters, 2-5 L, F/M Luer; 18/Pk

Representative image only.

MediaKap ME2M-050-18S Hollow Fiber Media Filters, 2-5 L, F/M Luer; 18/Pk

MediaKap | Mfr: ME2M-050-18S| Item: 2951002

Sterilize and clarify cell culture media and aqueous solutions quickly and efficiently

$1,608.94CAD / Box of 18

$1,608.94CAD / Box of 18
Product Description

Key Features

  • Filters may be used as pressure filters with peristaltic pumps, pressure vessels, or repeating syringes

More About this Item

MediaKap® Hollow Fiber Media Filter provides biocompatibility, low extractables, and high flow rates for serum-free media, and aqueous solutions. Polypropylene autovent prevents air locking. Naturally hydrophilic, 0.2 µm microporous membrane.

Sterile
Specifications
  • Membrane MaterialMixed Cellulose Ester
  • Pore Size (micron)0.2
  • Housing MaterialPolycarbonate
  • SterileYes
  • ConnectionsFemale luer by Male luer
  • Effective Filtration Area (cm²)70
  • Surface Area (m²)0.007
  • Typical Water Flow Rate750 mL/min at 10 psi
  • Max Operating Temperature (° C)83
  • Max Operating Temperature (° F)182
  • Max Pressure (PSI)50
  • Max Pressure3.45
  • Min Throughput (Liters)2
  • Max Throughput (Liters)5
  • Min Throughput (mL)2000
  • Max Throughput (mL)5000
  • DescriptionHollow Fiber Media Filters, 2-5 L, F/M Luer; 18/Pk
Reviews
Q&A