Mark-10 G1053 Universal V-Jaw Grip, Hex / #10-32F
Representative image only.
Mark-10 G1053 Universal V-Jaw Grip, Hex / #10-32F
Mark-10 | Mfr: G1053| Item: 5984837
Securely grips bottles and containers for closure torque testing
$581.99CAD / Each
$581.99CAD / Each
Product Description
Key Features
  • Torque capacity of 100 lbf in
  • Ideal for use with TST AND TSTM-DC
  • 3 year warranty
More About this Item
These grips are a key component of testing systems, designed for a wide range of torque applications.
This universal v-jaw grip, is ideal for gripping round objects. Serrated, reversible aluminum jaws effectively grip a wide range of sample shapes and sizes. Ergonomic, rubberized knob easily engages and disengages samples. Can be used either manually with an R50 torque sensor or TT03 torque gauge, or in test stand controlled applications with the TSTM-DC torque test stand.
3 Year Warranty
Specifications
  • DescriptionCap grip for 59848-00 and -02 test stands (0.65"-1.80" diameter)