Mark-10 Distance Measurement Package for Test Frames with EasyMESUR® Control Panel
Representative image only.
Mark-10 Distance Measurement Package for Test Frames with EasyMESUR® Control Panel
Mark-10 | Mfr: EMP001-1| Item: 6780114
Enhance your digital force tester with EasyMESUR® control panel with advanced applications
$2,983.61CAD / Each
$2,983.61CAD / Each
Product Description
Key Features
  • Field upgradeable—no need to ship it to the factory or call for service
  • Upgrade test frames 67801-10 through -13
  • Includes upgrades for distance measurement, distance limits, and preload
  • 3 year warranty
More About this Item
Bundled software packages offer convenience. Optional functions include, distance measurement and limits, speed extension, break detection, load holding, and cycling. Software can be added at any time and easily upgraded in the field.
Distance measurement: Measure distance with the ability to zero the reading.
Distance limits: Stop at user-defined upper and lower distance limits.
Preload: Zero the travel position automatically at a user-defined preload value.
Specifications
  • Compatible WithTest frames with EasyMESUR® control panel 67801-10 through -13
  • DescriptionDistance Measurement Package for Test Frames with EasyMESUR® Control Panel
  • Warranty3 years