Mark-10 AFCOMP Complete Options Package for ESM303
Representative image only.
Mark-10 AFCOMP Complete Options Package for ESM303
Mark-10 | Mfr: AFCOMP| Item: 2530333
Configurable for your application needs!
$6,689.19CAD / Each
$6,689.19CAD / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
This software options package for the Mark-10 ESM303 motorized push / pull test stand includes all functions listed below (25303-19 through -32), plus interface cable (25303-16) and USB cable (25303-17). Please see individual sku details or data sheet for more information on capabilities.
Requires a Series 7 or 5 gauge or indicator with load cell. Field hardware installation also required (if not ordered at the same time as the test stand).
Please note: The Mark-10 ESM303 motorized push / pull test stand and additional hardware are sold separately.
    Includes the follow software options:
    FollowMe™|Travel indication|Computer control|Programmable travel limits|Overload protection|Auto Return|Cycling/Dwell Time|Independent Up and Down Speeds|Low Speed Extension|High Speed Extension|Break Detection|Load Holding|Preload/Sample Touch|Profiles
Specifications
  • DescriptionPush / Pull Test Stand Complete Options Package