Mark-10 Advanced Functionality Package for Test Frames with EasyMESUR® Control Panel
Representative image only.
Mark-10 Advanced Functionality Package for Test Frames with EasyMESUR® Control Panel
Mark-10 | Mfr: EMP003-1| Item: 6780116
Enhance your digital force tester with EasyMESUR® control panel with advanced applications
$2,416.36CAD / Each
$2,416.36CAD / Each
Product Description
Key Features
  • Field upgradeable—no need to ship it to the factory or call for service
  • Upgrade test frames 67801-10 through -13
  • Includes upgrades for cycling, break detection, load holding, and FollowMe® function
  • 3 year warranty
More About this Item
Bundled software packages offer convenience. Optional functions include, distance measurement and limits, speed extension, break detection, load holding, and cycling. Software can be added at any time and easily upgraded in the field.
Cycling: Set up to 100,000 cycles, with user-defined upper and lower dwell times. Cycle between any combination of limit switches, travel limits, and load limits.
Break detection: Stop when a user-defined drop in force occurs.
Load holding: Stop at, and maintain, a user-defined load for a specified period of time. For best performance, use a force sensor with a capacity as close as possible to the target load.
FollowMe®: Push and pull on the sensor’s loading shaft to move the crosshead up and down, ideal for manual positioning prior to a test. Speed increases with applied force.
Specifications
  • Compatible WithTest frames with EasyMESUR® control panel 67801-10 through -13
  • DescriptionAdvanced Functionality Package for Test Frames with EasyMESUR® Control Panel
  • Warranty3 years