MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 10, 1 pair
Representative image only.
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 10, 1 pair
MAPA | Mfr: 6503110| Item: 8635743
Inner fabric lining provides exceptional comfort
$167.97CAD / Pair of 1
BUY MORE & SAVE
  • 1-5$167.97 CAD / Pair of 1
  • 6+$151.17 CAD / Pair of 1
$167.97CAD / Pair of 1
BUY MORE & SAVE
  • 1-5$167.97 CAD / Pair of 1
  • 6+$151.17 CAD / Pair of 1
Product Description
More About this Item
Butyl gloves provide excellent chemical resistance in applications involving acids, ketones, esters, and amine derivatives. 100% seamless cotton liner provides extra comfort for long-term use. Scalloped cuffs can be rolled down to prevent drips from reaching your arms. Unique Z-pattern grip design for optimal grip.
Specifications
  • Thickness (mil)60
  • MaterialButyl
  • Size10
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • LiningCotton
  • CuffScalloped
  • SterileNo
  • DescriptionCotton-Lined Butyl Gloves, size 10, 1 pair