MAPA 994928 Gloves, neoprene/nitrile/rubber, large
Representative image only.
MAPA 994928 Gloves, neoprene/nitrile/rubber, large
MAPA | Mfr: 34994018| Item: 4021502
Tri-polymer blend of nitrile, neoprene, and rubber
$52.75CAD / Bag of 100
BUY MORE & SAVE
  • 1-9$52.75 CAD / Bag of 100
  • 10+$47.48 CAD / Bag of 100
$52.75CAD / Bag of 100
BUY MORE & SAVE
  • 1-9$52.75 CAD / Bag of 100
  • 10+$47.48 CAD / Bag of 100
Product Description
Key Features
  • Tear resistance
  • Powder-free
More About this Item
Powder-free gloves provide added strength, tear resistance, and chemical splash protection. Longer 10" (25 cm) gloves provide added barrier protection.
Silicone free to prevent traces, defects on metal sheet and glass prior to painting. Ambidextrous.
Specifications
  • MaterialNitrile
  • Thickness (mil)6
  • SizeLarge
  • Length (in)10
  • Length (cm)25.4
  • ColorPurple
  • GripPebbled
  • CuffRolled
  • ComplianceFDA 21 CFR parts 170-199; EN ISO 374-1 Type B; EN ISO 374-5
  • SterileNo
  • DescriptionGloves, neoprene/nitrile/rubber, large