MadgeTech pHTemp2000 pH and Temperature Data Logger w/LCD
Representative image only.
MadgeTech pHTemp2000 pH and Temperature Data Logger w/LCD
MadgeTech | Mfr: PHTEMP2000| Item: 2300028
pHTemp2000
$1,181.66CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,181.66CAD / Each
Buy It Together
Current Item
MadgeTech pHTemp2000 pH and Temperature Data Logger w/LCD
MadgeTech pHTemp2000 pH and Temperature Data Logger w/LCD
2300028
$1,181.66CAD / Each
+ + =
Total Price 1879.31 ADD 3 ITEMS TO CART
Product Description
Includes
Data logger comes with power adapter.
Key Features
  • Large backlit LCD
  • Front panel keypad
  • Real-time operation
  • On-screen statistics
  • High-speed download
  • pH probe input jack
  • 1 year warranty
More About this Item
The pHTemp2000 is a pH and RTD temperature data recorder with LCD display. It accepts industry standard pH and ORP probes with female BNC connectors. The temperature compensation is provided through the external RTD temperature channel. It measures the full pH range from 0 to 14pH. The ultra-high input impedance (1012) of the pH2000 permits the use of standard high impedance probes.
The pHTemp2000 is ideal for applications such as waste water processing, environmental and wetland monitoring, surface and ground water quality monitoring. The device can be used with auto compensation (ATC) pH probes.
Canadian Standards Association1 Year Warranty
Specifications
  • Number of Channels2
  • Input TypeTemperature and pH
  • Max Temperature (° C)850
  • Min Temperature (° C)-200
  • Temperature Resolution (° C)0.01
  • Temperature Accuracy (° C)±0.1 (25 ambient)
  • Max pH14
  • Min pH0
  • Memory Size131,071/channel
  • Battery Life1 year
  • Length (in)4 51/64
  • Height (in)3 19/64
  • Width (in)1 1/4
  • DescriptionpH and Temperature Data Logger w/LCD