M-0890 Micro A07 Citric Acid Liquid Cleaner, 225 KG
Representative image only.
M-0890 Micro A07 Citric Acid Liquid Cleaner, 225 KG
International Products Corp | Mfr: M-0890-055| Item: 1810007
Environmentally friendly – Phosphate Free
$5,687.29CAD / Each
$5,687.29CAD / Each
Product Description
Key Features
  • Free Rinsing
More About this Item
Micro A07 is a free-rinsing cleaner designed for use in a wide range of applications including ultrasonic baths, immersion tanks, mild-agitation systems, CIP systems and more. It is ideal for cleaning metals, stainless steel, ceramics, glass, plastics, and filter membranes. Micro A07 solution removes a variety of soils as well including scale, oxides, milkstone, mineral deposits, and inorganic materials. It is free of CFCs, ODCs, phosphates, silicates, borates, halogenated compounds, and phenols.
Specifications
  • FormatLiquid
  • Dilution1:100 to 2:100, pH 3.0
  • ApplicationsInsoluble sales, Oxides
  • Description M-0890 MICRO A07 CITRIC ACID CLEANER 225 KG NET WT PLASTIC DRUM