Luxor Modular Room Divider Wall System, Add-On Wall, 70" x 48"; 2/Pk

Representative image only.

Luxor Modular Room Divider Wall System, Add-On Wall, 70" x 48"; 2/Pk

Luxor | Mfr: MW-7048-XFCG| Item: 4760085

Easily divide larger areas into quiet functional spaces

$1,019.59CAD / Pkg of 2

$1,019.59CAD / Pkg of 2
Product Description

Key Features

  • Dampen sound—PET dividers are rated to absorb 85% of ambient noise
  • Panels are constructed of a mix of PET and frosted acrylic to absorb noise without blocking natural light
  • Choose between starter and add-on walls to create a custom configuration to accommodate any workspace

More About this Item

The Luxor modular divider wall system is designed to divide larger areas into quieter, more functional spaces. The system features a mix of sound-absorbent PET and semi-transparent frosted acrylic inner panels that you can arrange to optimize privacy, reduce noise, and enhance natural light. The PET panels absorb 85% of ambient noise (NRC 0.85), are tackable for posting notes or pictures, and can be cleaned with up to 75% isopropyl alcohol. The frosted acrylic panels enhance privacy without blocking natural light, can double as a whiteboard when used with water-soluble dry-erase markers, and can be cleaned with mild soap and water.

Starter walls are freestanding and can be used as a stand-alone solution or connected to an add-on wall to create a custom configuration. Each wall features a built-in cable management tray to help keep cords organized, and its durable aluminum frame with sturdy steel feet to ensure stability and years of trouble-free use.

Specifications
  • Dimension48" H Add-On Wall
  • Width (in)70
  • Height (in)48
  • Depth (in)15 3/4
  • Width (cm)177.8
  • Height (cm)121.92
  • Depth (cm)40
  • MaterialAluminum frame, PET and frosted acrylic panels, powder-coated steel base and tray
  • Shipping weight (lbs)40
  • ColorGray PET panels and frosted acrylic panels
  • ComplianceNRC 0.85 Sound Absorption from PET Panels
  • DescriptionModular Room Divider Wall System, Add-On Wall, 70" x 48"; 2/Pk
Reviews
Q&A