Lovibond Color Meter for Surfaces, Liquids, and Powders
Representative image only.
Lovibond Color Meter for Surfaces, Liquids, and Powders
Lovibond | Mfr: 401120| Item: 5973450
Don't be limited to surface color measurements—unique stand lets you also measure powders, liquids, and pastes!
$11,352.47CAD / Each
$11,352.47CAD / Each
Buy It Together
Current Item
Lovibond Color Meter for Surfaces, Liquids, and Powders
Lovibond Color Meter for Surfaces, Liquids, and Powders
5973450
$11,352.47CAD / Each
+
Lovibond Disposable Cuvettes, 10-mm, 100/pack
5973452
$215.11CAD / Pkg of 100
=
Total Price 11567.58 ADD 2 ITEMS TO CART
Product Description
Includes
Stand with cuvette holder and cap, ten cuvettes, lanyard, protective pouch, carrying case, rechargeable lithium-ion battery, and USB cable.
Key Features
  • Attach text or an audio recording to your results for easy identification
  • Illuminate your sample using eight visible and one UV wavelength for more accurate color measurement
  • Each measurement is a combination of 27 images, giving information on color and texture effects
  • 1 year warranty
More About this Item
This handheld color meter measures surface color on graphic arts, painted surfaces, plastics, textiles, and more. Increase your application versatility by using the meter with its unique stand—enabling color measurements of liquids, pastes, and powders used in the food, cosmetics, and pharmaceutical industries.
The stand features a cuvette holder and secure cap to protect the sample from excessive ambient light. It also supports the meter at an ergonomic viewing angle.
The meter stores up to 350 sample measurements. Easily attach a text tag or audio message to any sample for information or identification purposes. Onboard software allows download of data in a formatted .pdf report or as a Microsoft® Excel® data table. Calibrate your color meter quickly and easily with the built-in reference slider.
1 Year Warranty
Specifications
  • Display Type1.75 in (4.5 cm) color display
  • Light Source25 LED array
  • Detector Type8 visible and 1 UV wavelength
  • Wavelength(s) (nm)0.10 delta E 94 on white
  • Memory Size350 sample measurements
  • OutputUSB
  • Battery1 Rechargeable Lithium-ion (included)
  • DescriptionColor Meter for Surfaces, Liquids, and Powders