Lovibond Color Meter for Surfaces, 4/8-mm measuring area
Representative image only.
Lovibond Color Meter for Surfaces, 4/8-mm measuring area
Lovibond | Mfr: 401100| Item: 5973451
Attach text or an audio recording to your results for easy identification
$6,843.90CAD / Each
$6,843.90CAD / Each
Product Description
Includes
Lanyard, protective pouch, carrying case, rechargeable lithium-ion battery, and USB cable.
Key Features
  • Illuminate your sample using eight visible and one UV wavelength for more accurate color measurement
  • Each measurement is a combination of 27 images, giving information on color and texture effects
  • 1 year warranty
More About this Item
This handheld color meter measures surface color on graphic arts, painted surfaces, plastics, textiles, and more. Compact size and rechargeable batteries let you use this unit in the lab or carry on the plant floor for quality control testing.
Store up to 350 sample measurements; attach a text tag or audio message to any sample for information or identification purposes. Onboard software allows download of data in a formatted report or as a Microsoft® Excel® data table. Calibrate your color meter quickly and easily with the built-in reference slider.
1 Year Warranty
Specifications
  • Display Type1.75 in (4.5 cm) color display
  • Light Source25 LED array
  • OutputUSB
  • Battery1 Rechargeable Lithium-ion (included)
  • DescriptionColor Meter for Surfaces, 4/8-mm measuring area