Lovibond AF228 Gardner Benchtop Gardner Comparator
Representative image only.
Lovibond AF228 Gardner Benchtop Gardner Comparator
Lovibond | Mfr: 345010| Item: 9956238
Economical, visual color measurement with three-field determination
$10,151.80CAD / Each
$10,151.80CAD / Each
Product Description
Includes
discs containing colored glass filters, sample cell/tubes, calibration filter with certificate, and power cord.
Key Features
  • Available with the most popular color scales
  • Quick comparison of sample to glass color standards
  • 1 year warranty
More About this Item
Convenient benchtop comparator is a three-field instrument for visually determining the color of samples by direct comparison with colored glass filters. With a 3-section field of view, the sample and two consecutive glasses on the color scale are viewed simultaneously, making it easier to achieve the optimal color match. For rapid color grading within predetermined color limits, the glasses can be set to the two limiting colors, making it easy to see if the sample is within tolerance.
1 Year Warranty
Specifications
  • TestChemicals, edible oils and fats, petroleum oils, waxes
  • DescriptionBenchtop Gardner Comparator