-Lite Resistivity Indicators, 1 m-ohm, 120 VAC
Representative image only.
-Lite Resistivity Indicators, 1 m-ohm, 120 VAC
Thermo Scientific| Item: 0150437
A low-cost alternative to digital purity meters
$394.50CAD / Each
$394.50CAD / Each
Product Description
Key Features
  • Designed for use with B-Pure systems
  • 1 year warranty
More About this Item
These indicators can be mounted directly on top of single- or doublecartridge housings with 1⁄2" NPT(F) connections.
White light indicates water resistivity is above the threshold; red light warns if resistivity falls below threshold. Operates on 120 VAC power.
1 Year Warranty
Specifications
  • Inlet Port1/2" NPT(F)
  • Resistivity Threshold200 kΩ
  • Description-Lite Resistivity Indicators, 1 m-ohm, 120 VAC