Lascar Electronics EL-USB-5+ Event, State Change and Counter Data Logger, Extended Memory
Representative image only.
Lascar Electronics EL-USB-5+ Event, State Change and Counter Data Logger, Extended Memory
Lascar Electronics | Mfr: EL-USB-5+| Item: 1800036
Download directly to PC via built-in USB
$206.27CAD / Each
$206.27CAD / Each
Product Description
Includes
configuration software and battery.
Key Features
  • Programmable alarms alert to problems
  • Easily manipulate and analyze data with included software
  • IP67-rated housing protects logger from soil and moisture (cap must be fitted)
  • 1 year warranty
More About this Item
—Rising voltage, falling voltage, or volt-free trigger options
—Log up to 2 state changes/second or 4 events/second
1 Year WarrantyConformite Europeenne
Specifications
  • Number of Channels1
  • Input TypeVoltage
  • Max Frequency (Hz)100
  • Min Frequency (Hz)0
  • Sampling Rate0 to 2 Hz (event/state change) or 0 to 100 Hz (count frequency)
  • Memory Size1007616
  • InterfaceSoftware
  • ConnectionsUSB
  • Battery Life1 year
  • Battery Type1 1/2 AA lithium (included)
  • DescriptionExtended memory event, state change & counter data logger