Lascar Electronics EL-USB-1 USB Temperature Data Logger, 16,000+ readings, 1 year battery life
Representative image only.
Lascar Electronics EL-USB-1 USB Temperature Data Logger, 16,000+ readings, 1 year battery life
Lascar Electronics | Mfr: EL-USB-1| Item: 2303950
Direct-to-USB connection and easy-to-use functionality
$107.70CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$107.70CAD / Each
Buy It Together
Current Item
Lascar Electronics EL-USB-1 USB Temperature Data Logger, 16,000+ readings, 1 year battery life
Lascar Electronics EL-USB-1 USB Temperature Data Logger, 16,000+ readings, 1 year battery life
2303950
$107.70CAD / Each
+ + =
Total Price 649.91 ADD 3 ITEMS TO CART
Product Description
Includes
configuration software and battery.
Key Features
  • User-programmable alarm thresholds — perfect for compliance monitoring
  • All data loggers come with software
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
For ambient temperature measurements between –31 to 176°F (–35 to 80°C), model EL-USB-1 stores over 16,000 readings.
IP 671 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature (one internal)
  • Max Temperature (° F)176
  • Min Temperature (° F)-31
  • Max Temperature (° C)80
  • Min Temperature (° C)-35
  • Temperature Resolution (° F)0.1
  • Temperature Accuracy (° F)±2
  • Temperature Resolution (° C)0.5
  • Temperature Accuracy (° C)±1
  • Sampling Rate1 per 10 seconds to 1 per 12 hours
  • Memory Size16,382 data points
  • Battery Life3 years (typical)
  • Battery Type1 lithium 1/2 AA (included)
  • Display TypeNo
  • Length (in)3
  • Width (in)1
  • DescriptionUSB Temperature Data Logger, 16,000+ readings, 1 year battery life