Lascar Electronics EL-GFX-TC USB Data Logger with Graphic LCD Screen, single thermocouple
Representative image only.
Lascar Electronics EL-GFX-TC USB Data Logger with Graphic LCD Screen, single thermocouple
Lascar Electronics | Mfr: EL-GFX-TC| Item: 2303911
Graphic LCD provides real-time information on data being recorded
$325.62CAD / Each
$325.62CAD / Each
Product Description
Includes
cover, micro USB cable, magnetic wall-mounting clip, and two 3.6 V lithium batteries. Models 23039-11 and 23039-13 include a temperature probe. Models 23039-06 and 23039-12 include two temperature probes.
Key Features
  • Ability to start, stop, graph trends, and summarize data in the field without computer connection
  • Memory capacity exceeds 250,000 readings per channel with resolution to the nearest 0.1 digit
  • Windows®-based control software available as free download
  • Compact and robust waterproof design with IP67 rating
  • Wall-mounting clip for quick magnetic mounting to a metallic surface
  • 1 year warranty
1 Year WarrantyConformite Europeenne
Specifications
  • Number of Channels1
  • Input TypeTemperature (thermocouple)
  • Display TypeLCD
  • DescriptionUSB Data Logger with Graphic LCD Screen, single thermocouple