Lascar Electronics EL-GFX-DTP USB Data Logger with Graphic LCD Screen, dual thermistor
Representative image only.
Lascar Electronics EL-GFX-DTP USB Data Logger with Graphic LCD Screen, dual thermistor
Lascar Electronics | Mfr: EL-GFX-DTP| Item: 2303912
Graphic LCD provides real-time information on data being recorded
$390.82CAD / Each
$390.82CAD / Each
Product Description
Includes
cover, micro USB cable, magnetic wall-mounting clip, and two 3.6 V lithium batteries. Models 23039-11 and 23039-13 include a temperature probe. Models 23039-06 and 23039-12 include two temperature probes.
Key Features
  • Ability to start, stop, graph trends, and summarize data in the field without computer connection
  • Memory capacity exceeds 250,000 readings per channel with resolution to the nearest 0.1 digit
  • Windows®-based control software available as free download
  • Compact and robust waterproof design with IP67 rating
  • Wall-mounting clip for quick magnetic mounting to a metallic surface
  • 1 year warranty
1 Year WarrantyConformite Europeenne
Specifications
  • Number of Channels2
  • Input TypeTemperature (two external)
  • Max Temperature (° F)257
  • Min Temperature (° F)-40
  • Max Temperature (° C)125
  • Min Temperature (° C)-40
  • Display TypeLCD
  • DescriptionUSB Data Logger with Graphic LCD Screen, dual thermistor