LaMotte P-6740-H Free Chlorine Test Reagent, DPD #1A; 60 Ml

Representative image only.

LaMotte P-6740-H Free Chlorine Test Reagent, DPD #1A; 60 Ml

LaMotte | Mfr: P-6740-H| Item: 0556428

Optimize performance of your LaMotte® colorimeter and turbidity meters

$38.68CAD / Each

$38.68CAD / Each

Buy It Together

Current Item

LaMotte P-6740-H Free Chlorine Test Reagent, DPD #1A; 60 Ml
LaMotte P-6740-H Free Chlorine Test Reagent, DPD #1A; 60 Ml
0556428

$38.68CAD / Each
+ =
Total Price 124.28 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Replacement parts, accessories, and reagents to keep your colorimeter and turbidity results dependable
  • Liquid alternative to DPD tablets

More About this Item

Choose from a wide range of accessories and reagents to measure water testing parameters utilizing LaMotte® colorimeter and turbidity meters. Meters sold separately.

DPD 1A and DPD 1B are added to a 5 or 10 mL sample to test free available chlorine. DPD 3 is added to the reacted sample to measure total chlorine.

Specifications
  • Test ParametersChlorine; Free
  • DescriptionFree Chlorine Test Reagent, DPD #1A; 60 mL
Reviews
Q&A