Labmat F246750020 Liner Sheets, 20 x 24"; 100 Sheets/Case

Representative image only.

Labmat F246750020 Liner Sheets, 20 x 24"; 100 Sheets/Case

Labmat | Mfr: F246750020| Item: 0680505

Skid resistent polyethylene film backing that prevents leakage

$521.58CAD / Case of 100

$521.58CAD / Case of 100
Product Description

Key Features

  • Disposable

More About this Item

Highly abosrbent top layer soaks up corrosive materials, and most liquids. Red backing alerts end-users when biohazard materials are in use. Liners can easily be cut to meet end users specific applications.

Specifications
  • MaterialPolyethylene
  • Width (in)20
  • Length (ft)2
  • Width (cm)50.8
  • Length (m)0.6096
  • ColorWhite
  • DescriptionLiner Sheets, 20 x 24"; 100 Sheets/Case
Reviews
Q&A