label 1.28125 in W x 0.28125 in H
Representative image only.
label 1.28125 in W x 0.28125 in H
| Item: 6220600
$115.51CAD / Roll of 1000
BUY MORE & SAVE
  • 1-5$115.51 CAD / Roll of 1000
  • 6-29$103.96 CAD / Roll of 1000
  • 30-119$98.18 CAD / Roll of 1000
  • 120+$92.41 CAD / Roll of 1000
$115.51CAD / Roll of 1000
BUY MORE & SAVE
  • 1-5$115.51 CAD / Roll of 1000
  • 6-29$103.96 CAD / Roll of 1000
  • 30-119$98.18 CAD / Roll of 1000
  • 120+$92.41 CAD / Roll of 1000
Product Description
Key Features
  • 1 year warranty
More About this Item
Cryo-Babies cryogenic labels are constructed of flexible polyvinyl and feature chemical-resistantce. Labels withstand temperatures down to -196°C and adhere to plastics, glass, and metal. Mark using any writing instrument. Suitable for autoclaving, boiling, water baths, or dry heat up to 150°C without peeling, cracking, or degrading. Lables can survive soaking at room temperature or 100% relative humidity at 35°C for at least 24 hours.
1 Year Warranty
Specifications
  • ColorWhite
  • Length (in)1.2812
  • Width (in)0.2812
  • Descriptionlabel 1.28125 in W x 0.28125 in H