Labconco Stobe Light for CentriVap Concentrator
Representative image only.
Labconco Stobe Light for CentriVap Concentrator
Labconco | Mfr: 7464300| Item: 2865660
Optimize performance of your CentriVap concentrator
$1,186.08CAD / Each
$1,186.08CAD / Each
Product Description
Key Features
  • Suitable for use with all CentriVap benchtop and mobile system concentrators
  • 1 year warranty
More About this Item
CentriZap Strobe Light enables the user to shine a strobe light into the CentriVap's chamber to easily view samples while the rotor is spinning. Includes mounting holster and hardware for attachment to the right side of the CentriVap. CentriVap not included.
1 Year Warranty
Specifications
  • DescriptionStobe Light for CentriVap Concentrator