Labconco Purifier 3980423 Class 1 Filtered Enclosure, 4' W, 230 VAC

Representative image only.

Labconco Purifier 3980423 Class 1 Filtered Enclosure, 4' W, 230 VAC

Labconco Purifier | Mfr: 3980423| Item: 3350710

Protect you from biological hazards requiring Biosafety Level 1, 2, and 3 containment

$15,949.45CAD / Each

$15,949.45CAD / Each
Product Description

Key Features

  • EPA filter removes 99.99% of particulates down to 0.3 µm
  • 1 year warranty

More About this Item

Purifier enclosures protect you and your laboratory environment—they offer an economical alternative to Class II laminar flow cabinets when your applications do not require product protection. The filter indicator monitors the differential pressure across the filter and lets you know when to change the filter. Side panels and pivoting, angled sash are made of tempered safety glass; sash pivots upward 20" for easy cabinet loading. All units are shipped fully assembled. Order a spill-containing work surface and other accessories separately.

Class I Enclosures protect the user from agents that require Biosafety Level 1, 2, or 3 containment but no product protection. These enclosures include an ultraviolet light that may be used in conjunction with surface disinfection to ensure thorough decontamination.

1 Year WarrantyETL ListedISO9001:2000 Certified Supplier
Specifications
  • Hood Width (in)48
  • Blower TypeStandard
  • LightsUV
  • Exterior Width (in)47.75
  • Exterior Height (in)37.0625
  • Exterior Depth (in)28.6562
  • Power (VAC)230
  • Power (Hz)50
  • Exhaust Volume (CFM)220 to 310
  • DescriptionClass I Filtered Enclosure, 4' W, 230V
  • Warranty1 Year
Reviews
Q&A