Labconco Protector® Replacement Acid Roughing Prefilter for Protector® Echo™ and Airo™ Fume Hoods, after 2020
Representative image only.
Labconco Protector® Replacement Acid Roughing Prefilter for Protector® Echo™ and Airo™ Fume Hoods, after 2020
Labconco | Mfr: 9577423| Item: 2861712
Optimize performance of your Labconco Protector® Echo™ ductless fume hoods
$231.32CAD / Each
$231.32CAD / Each
Product Description
Includes
26617, 286170, 28617 0, 28617-0, 286171, 28617 1, 28617-1, filter, prefilter, HEPA, Neutrodine, Unisorb, Molecular
Key Features
  • Prefilter, filter, and HEPA filters ensure user safety
  • Compatible with Labconco® Protector® Echo™ and Airo™ ductless fume hoods
  • Treated for use with acids
  • 1 year warranty
More About this Item
For use with Protector Echo and Airo Filtered Fume Hoods manufactured after September 2020.
1 Year Warranty
Specifications
  • Filter TypeAcid roughing
  • DescriptionReplacement Acid Roughing Prefilter for Protector® Echo™ and Airo™ Fume Hoods, after 2020