Labconco Protector® HEPA Filter for Protector® Echo™ and Airo™ Fume Hoods, 9/2020 to 2/2025
Representative image only.
Labconco Protector® HEPA Filter for Protector® Echo™ and Airo™ Fume Hoods, 9/2020 to 2/2025
Labconco | Mfr: 9577421| Item: 2861710
Optimize performance of your Labconco Protector® Echo™ ductless fume hoods
$1,458.66CAD / Each
$1,458.66CAD / Each
Product Description
Includes
26617, 286170, 28617 0, 28617-0, 286171, 28617 1, 28617-1, filter, prefilter, HEPA, Neutrodine, Unisorb, Molecular
Key Features
  • Prefilter, filter, and HEPA filters ensure user safety
  • Compatible with Labconco® Protector® Echo™ and Airo™ ductless fume hoods
  • 99.995% efficient
  • H14 type
  • 1 year warranty
More About this Item
For use with Protector Echo and Airo Filtered Fume Hoods manufactured after September 2020 and before February 2025.
1 Year Warranty
Specifications
  • Filter TypeHEPA (for models from 9/20 to 2/25)
  • DescriptionHEPA Filter for Protector® Echo™ and Airo™ Fume Hoods, 9/2020 to 2/2025