Labconco Neutrodine Molecular Filter for Protector® Echo™ and Airo™ Fume Hoods
Representative image only.
Labconco Neutrodine Molecular Filter for Protector® Echo™ and Airo™ Fume Hoods
Labconco | Mfr: 9577400| Item: 3253500
Optimize performance of your Labconco Protector® Echo™ ductless fume hoods
$2,460.56CAD / Each
$2,460.56CAD / Each
Product Description
Includes
26617, 286170, 28617 0, 28617-0, 286171, 28617 1, 28617-1, filter, prefilter, HEPA, Neutrodine, Unisorb, Molecular
Key Features
  • Prefilter, filter, and HEPA filters ensure user safety
  • Compatible with Labconco® Protector® Echo™ and Airo™ ductless fume hoods
  • For multi-purpose use of acids/bases/solvents
  • 1 year warranty
More About this Item
For use with Protector Echo and Airo Ductless Fume Hoods manufactured before September 2020.
1 Year Warranty
Specifications
  • Filter TypeNeutrodine molecular
  • DescriptionNeutrodine Molecular Filter for Protector® Echo™ and Airo™ Fume Hoods