Labconco FreeZone Standard Freeze Drying Chamber, 16 Ports, SS
Representative image only.
Labconco FreeZone Standard Freeze Drying Chamber, 16 Ports, SS
Labconco | Mfr: 7522900| Item: 0334611
Easily attach flasks to your freeze dry system or contain samples in bulk
$6,048.27CAD / Each
$6,048.27CAD / Each
Buy It Together
Current Item
Labconco FreeZone Standard Freeze Drying Chamber, 16 Ports, SS
Labconco FreeZone Standard Freeze Drying Chamber, 16 Ports, SS
0334611
$6,048.27CAD / Each
+ + + =
Total Price 21078.32 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Sixteen neoprene valves and plastic knobs.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Accommodates ½” and ¾” adapters
  • Compatible with 4.5-, 6-, 12-, and 18-L FreeZone Freeze Dryers
  • 1 year warranty
More About this Item
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk. Various models are available from 8-, 12- to 16-port drying chambers to 3 to 5 heated product shelf chambers.
The stainless-steel drying chamber features 16 neoprene valves with plastic knobs that accommodate both ½” and ¾” adapters for quick and easy connection of flasks.
1 Year Warranty
Specifications
  • Exterior Height (InchShort)13
  • DescriptionStandard Freeze Drying Chamber, 16 Ports, SS