Labconco 9410300 Sash Stop for Protector Premier Hoods
Representative image only.
Labconco 9410300 Sash Stop for Protector Premier Hoods
Labconco | Mfr: 9410300| Item: 0907084
Restricts how far a vertical-rising sash may be opened
$94.59CAD / Each
$94.59CAD / Each
Product Description
Key Features
  • Easy to install on fixture panel
  • 1 year warranty
More About this Item
Sash stop restricts the sash opening height of your fume hood. With the sash kept in a lowered position, the exhaust volume demand on the blower is reduced. Release the sash stop to open the sash completely for easier hood loading and cleaning. Sash stop kit can be used on most Labconco fume hoods made since 2012.
1 Year Warranty
Specifications
  • DescriptionSash Stop for Protector Premier Hoods