Labconco 9041300 WaterPro BT Replacement UV Lamp, 185/254 nm; 1/Pk
Representative image only.
Labconco 9041300 WaterPro BT Replacement UV Lamp, 185/254 nm; 1/Pk
Labconco | Mfr: 9041300| Item: 9929051
Optimize performance of your Labconco WaterPro BT Water Purification Systems
$1,362.89CAD / Each
$1,362.89CAD / Each
Product Description
Key Features
  • Compatible with the Labconco WaterPro BT Water Purification Systems
  • Compatible with WaterPro BT Systems that came installed with the UV Lamp option only
  • 1 year warranty
More About this Item
Choose from a wide range of accessories and replacement parts for your Labconco WaterPro BT Water Purification Systems.
Replacement UV lamp/bulb operates at 185 and 254 nm to remove bacteria, their metabolites, and certain organics.
1 Year Warranty
Specifications
  • DescriptionWaterPro BT Replacement UV Lamp, 185/254 nm; 1/Pk