Labconco 7553001 Acid-Resistant 6-Place Microtiter Plate Rotor
Representative image only.
Labconco 7553001 Acid-Resistant 6-Place Microtiter Plate Rotor
Labconco | Mfr: 7553001| Item: 1304060
Enhance the performance of your Labconco CentriVap® Vacuum Concentrator with these accessories
$5,105.30CAD / Each
$5,105.30CAD / Each
Product Description
Key Features
  • Choose replacement parts or optional accessories based on your application needs
  • Accommodates up to 6 standard 96 well plates or 3 deep well plates
  • PTFE coating provides protection against inert chemicals and harsh applications
  • Six place microtiter plate makes it easy to interchange rotors
More About this Item
Choose from a wide range of rotors and other accessories suitable for use with your Labconco CentriVap® Complete™ Vacuum Concentrator.
Specifications
  • Capacity (Gal)6-standard 96 standard or 3 deep well plates
  • MaterialPTFE-coated anodized aluminum, PP hub
  • Compatible WithCentriVap® Centrifugal Vacuum Concentrator systems
  • DescriptionAcid-Resistant 6-Place Microtiter Plate Rotor