Labconco 5005800 Glove Box Sleeves, Straight, 28" Long with Hands, Neoprene; 1 Pair
Representative image only.
Labconco 5005800 Glove Box Sleeves, Straight, 28" Long with Hands, Neoprene; 1 Pair
Labconco | Mfr: 5005800| Item: 0911218
Ideal for use with neoprene or latex glove box hands
$920.87CAD / Pair of 1
$920.87CAD / Pair of 1
Product Description
Includes
O-rings and a spring.
Key Features
  • 1 year warranty
More About this Item
These neoprene straight sleeves are ideal for use with neoprene or latex glove box hands. The sleeves fit 8 to 8-3/4" openings.
Order glove box hands separately.
1 Year Warranty
Specifications
  • Transfer Chamber Length (in)28
  • DescriptionGlove Box Sleeves, Straight, 28" Long with Hands, Neoprene; 1 Pair