Labconco 3955202 Single-Filter Ammonia Fume Adsorber, 115V

Representative image only.

Labconco 3955202 Single-Filter Ammonia Fume Adsorber, 115V

Labconco | Mfr: 3955202| Item: 0900360

Trap harmful xylene and toluene fumes

$5,429.44CAD / Each

$5,429.44CAD / Each

Buy It Together

Current Item

Labconco 3955202 Single-Filter Ammonia Fume Adsorber, 115V
Labconco 3955202 Single-Filter Ammonia Fume Adsorber, 115V
0900360

$5,429.44CAD / Each
+ =
Total Price 6423.98 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

Fume adsorbers are designed for light-duty applications involving organic solvents, formaldehyde, ammonia, or low molecular weight amines. Fume adsorbers use carbon filtration to trap vapors so that safe, filtered air is returned to the laboratory. No outside ducting is required. These portable, low-profile enclosures fit easily on countertops with overhanging cabinets and are available in two widths.
Each fume adsorber includes carbon filters that adsorbs vapors from organic solvents, formaldehyde or ammonia. Models 09003-40, -45, -75, -80, -85 and -90 require two filters; all other models require one filter. Features include built-in motorized impeller to provide face velocity up to 35 fpm. Adsorber has a white, dry powder epoxy-coated steel housing with perforated baffle for uniform airflow. Canopy is a clear, one-piece 3/8"-thick, shatter-resistant acrylic. Quiet operation is 58 to 63 dBA. A pre-wired rear-mounted auxiliary electrical receptacle has a top-mounted blower switch with circuit breaker. Work surface and base cabinet must be ordered separately.
What's included: Carbon filter (two for double models), power switch, and an 8-ft cord. The 115 VAC models include a three-prong plug.

1 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Specifications
  • Switches1
  • Exterior Width (in)30
  • Exterior Height (in)15.5
  • Exterior Depth (in)23.5
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionSingle-Filter Ammonia Fume Adsorber, 115V
  • Warranty1 year
Reviews
Q&A