Labconco 3937302 Trace Odor Ammonia & Amines Carbon Filter for 3-ft & 6-ft Xpert® Filtered Systems (2 Required for 6-ft System)
Representative image only.
Labconco 3937302 Trace Odor Ammonia & Amines Carbon Filter for 3-ft & 6-ft Xpert® Filtered Systems (2 Required for 6-ft System)
Labconco | Mfr: 3937302| Item: 0910973
Trap nonvolatile nuisance odors and vapors from samples
$1,296.58CAD / Each
$1,296.58CAD / Each
Product Description
Key Features
  • Carbon filters traps odors and minute quantities of chemicals
  • Compatible with 3-ft and 6-ft Xpert filtered systems, 6-ft required 2 filters
  • 9.2 lb activated carbon
More About this Item
Easily install atop the XPert filtered systems to trap odors and small quantities of chemical vapors.
Conformite EuropeenneETL ListedISO9001:2000 Certified Supplier
Specifications
  • Compatible With3-ft & 6-ft Xpert® filtered systems (2 required for 6-ft system)
  • TypeAmmonia & amines carbon filter
  • DescriptionTrace Odor Ammonia & Amines Carbon Filter for 3-ft & 6-ft Xpert® Filtered Systems (2 Required for 6-ft System)