Lab-Crest 110-601-0001 Female Coupling Component, Aluminum
Representative image only.
Lab-Crest 110-601-0001 Female Coupling Component, Aluminum
Lab-Crest | Mfr: 110-601-0001| Item: 6340064
Optimize performance of your Lab-Crest glassware
Call For Price
Call For Price
Buy It Together
Current Item
Lab-Crest 110-601-0001 Female Coupling Component, Aluminum
Lab-Crest 110-601-0001 Female Coupling Component, Aluminum
6340064
Call For Price
+ +
Lab-Crest 110-650-0001 Male Coupling Component, SS
6340066
$222.48CAD / Each
=
Total Price 594.88 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Aluminum construction
More About this Item
A complete coupling consists of a female coupling (63400-64 or 63400-72), male coupling (63400-66), insert (63400-68 or 63400-78), and an O-ring.
Specifications
  • MaterialAluminum
  • Compatible WithPressure reaction vessels
  • TypeFemale Coupling
  • DescriptionFemale Coupling Component, Aluminum