Koflo 3/4-SA-3-12R-11 Sanitary Mixer, 3/4" Tri-clamp size, 12 elements
Representative image only.
Koflo 3/4-SA-3-12R-11 Sanitary Mixer, 3/4" Tri-clamp size, 12 elements
Koflo | Mfr: 3/4-SA-3-12R-11| Item: 0466834
$4,652.97CAD / Each
$4,652.97CAD / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Blade Design Mixers are best for low-viscosity (<1,500 Cp) fluids. The faster the fluid moves, the faster (and more thorough) the mixing. Great for mixing products into water or water-like fluids.
Limited Lifetime Warranty
Specifications
  • Material316 Stainless Steel
  • No. Of Elements12
  • Pipe Id (in)0.62
  • Pipe Id (mm)15.75
  • Mixer Length (in)13
  • Mounting Type0.75" Tri-Clamp
  • DescriptionSanitary Mixer, 3/4" Tri-clamp size, 12 elements
  • WarrantyLTD Life Time