Koflo 2.5-SA-3-6R-11 Sanitary Mixer, 2 1/2" Tri-clamp size, 6 elements
Representative image only.
Koflo 2.5-SA-3-6R-11 Sanitary Mixer, 2 1/2" Tri-clamp size, 6 elements
Koflo | Mfr: 2.5-SA-3-6R-11| Item: 0466848
$5,598.88CAD / Each
$5,598.88CAD / Each
Product Description
Key Features
  • Limited Lifetime Warranty
Limited Lifetime Warranty
Specifications
  • Material316 Stainless Steel
  • No. Of Elements6
  • Pipe Id (in)2.37
  • Pipe Id (mm)60.2
  • Mixer Length (in)19
  • Mounting Type2.5" Tri-Clamp
  • DescriptionSanitary Mixer, 2 1/2" Tri-clamp size, 6 elements
  • WarrantyLTD Life Time