Koflo 1.5-SA-3-6R-11 Sanitary Mixer, 1 1/2" Tri-clamp size, 6 elements
Representative image only.
Koflo 1.5-SA-3-6R-11 Sanitary Mixer, 1 1/2" Tri-clamp size, 6 elements
Koflo | Mfr: 1.5-SA-3-6R-11| Item: 0466840
$4,104.86CAD / Each
$4,104.86CAD / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Blade Design Mixers are best for low-viscosity (<1,500 Cp) fluids. The faster the fluid moves, the faster (and more thorough) the mixing. Great for mixing products into water or water-like fluids.
Limited Lifetime Warranty
Specifications
  • Material316 Stainless Steel
  • No. Of Elements6
  • Pipe Id (in)1.37
  • Pipe Id (mm)34.8
  • Mixer Length (in)12
  • Mounting Type1.5" Tri-Clamp
  • DescriptionSanitary Mixer, 1 1/2" Tri-clamp size, 6 elements
  • WarrantyLTD Life Time