KNF UN726FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 0.6 cfm/27.95"Hg-20psi

Representative image only.

KNF UN726FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 0.6 cfm/27.95"Hg-20psi

KNF | Mfr: 073258/075636| Item: 0705761

Oversized, permanently lubricated bearings for long-term operation

$3,841.13CAD / Each

$3,841.13CAD / Each

Buy It Together

Current Item

KNF UN726FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 0.6 cfm/27.95"Hg-20psi
KNF UN726FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 0.6 cfm/27.95"Hg-20psi
0705761

$3,841.13CAD / Each
+ =
Total Price 11745.87 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

More About this Item

Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system.

Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.

UL Listed to Canadian safety standards.UL Listed.1 Year Warranty
Specifications
  • Head ConfigurationDual Heads
  • Wetted PartsPTFE
  • Free-Air Capacity (CFM)0.6
  • Free-Air Capacity (LPM)17
  • Free-Air Capacity (LPH)1020
  • Max Flow Rate (CFM)0.6
  • Max Flow Rate (Liters/min)17
  • Max Vacuum (in Hg)27.95
  • Max Vacuum (mbar)66.8
  • Max Vacuum (mm Hg)50.1
  • Max Vacuum (torr)50.1
  • Port Size1/4" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115 / 230
  • Power (amps)1.9
  • Power (Hz)50/60
  • Height (in)10.875
  • Depth (in)12.875
  • Width (in)10.72
  • Height (cm)27.62
  • Depth (cm)32.7
  • Width (cm)27.23
  • Height (mm)276.2
  • Depth (mm)327
  • Width (mm)272.3
  • DescriptionExplosion Proof Motor Vacuum Pump, PTFE/PTFE; 0.6 cfm/27.95"Hg-20psi
  • Warranty1 Year
Reviews
Q&A