KNF UN726.1.2FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 1.1 cfm/27.95"Hg-20psi

Representative image only.

KNF UN726.1.2FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 1.1 cfm/27.95"Hg-20psi

KNF | Mfr: 073291/076071| Item: 0705762

Oversized, permanently lubricated bearings for long-term operation

$4,707.48CAD / Each

$4,707.48CAD / Each

Buy It Together

Current Item

KNF UN726.1.2FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 1.1 cfm/27.95"Hg-20psi
KNF UN726.1.2FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 1.1 cfm/27.95"Hg-20psi
0705762

$4,707.48CAD / Each
+ =
Total Price 12612.22 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

More About this Item

Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system.

Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.

1 Year Warranty
Specifications
  • Head ConfigurationDual Heads
  • Wetted PartsPTFE
  • Free-Air Capacity (CFM)1.1
  • Free-Air Capacity (LPM)31.1
  • Free-Air Capacity (LPH)1866
  • Max Flow Rate (CFM)1.05
  • Max Flow Rate (Liters/min)30
  • Max Vacuum (in Hg)27.95
  • Max Vacuum (mbar)66.8
  • Max Vacuum (mm Hg)50.1
  • Max Vacuum (torr)50.1
  • Port Size1/4" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115 / 230
  • Power (amps)2
  • Power (Hz)50/60
  • Height (in)10.875
  • Width (in)8.3125
  • Length (in)13.6875
  • Height (cm)27.62
  • Width (cm)21.11
  • Length (cm)34.77
  • Height (mm)276.2
  • Width (mm)211.1
  • Length (mm)347.7
  • DescriptionExplosion Proof Motor Vacuum Pump, PTFE/PTFE; 1.1 cfm/27.95"Hg-20psi
  • Warranty1 Year
Reviews
Q&A