KNF UN026STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 0.6 cfm/26.9"Hg-36psi

Representative image only.

KNF UN026STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 0.6 cfm/26.9"Hg-36psi

KNF | Mfr: 073301/075636| Item: 0705771

Oversized, permanently lubricated bearings for long-term operation

$3,810.19CAD / Each

$3,810.19CAD / Each

Buy It Together

Current Item

KNF UN026STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 0.6 cfm/26.9"Hg-36psi
KNF UN026STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 0.6 cfm/26.9"Hg-36psi
0705771

$3,810.19CAD / Each
+ =
Total Price 11714.93 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

More About this Item

Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system.

Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.

1 Year Warranty
Specifications
  • Head ConfigurationSingle Head
  • Wetted Parts316 Stainless Steel, PTFE
  • Free-Air Capacity (CFM)0.6
  • Free-Air Capacity (LPM)17
  • Free-Air Capacity (LPH)1020
  • Max Flow Rate (CFM)0.6
  • Max Flow Rate (Liters/min)17
  • Max Vacuum (in Hg)26.9
  • Max Vacuum (mbar)102.4
  • Max Vacuum (mm Hg)76.7
  • Max Vacuum (torr)76.7
  • Port Size1/4" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115 / 230
  • Power (amps)2.1
  • Power (Hz)50/60
  • Height (in)10.9375
  • Width (in)5.8125
  • Length (in)12.9375
  • Height (cm)27.78
  • Width (cm)14.76
  • Length (cm)32.86
  • Height (mm)277.8
  • Width (mm)147.6
  • Length (mm)328.6
  • DescriptionExplosion Proof Motor Vacuum Pump, 316SS/PTFE; 0.6 cfm/26.9"Hg-36psi
  • Warranty1 Year
Reviews
Q&A